Learn More
SymLine™ System Accessories
Product Code. 17337419
Click to view available options
Product Type:
Blind Plug
Cabinet Feed-through
Grounding Cable
Mounting Bracket
Color:
Black
Silver
Yellow
For Use With (Equipment):
32 mm Dia. Pipe
Cabinet (28 mm Borehole, 120 mm Wall Thickness)
Pipe System
SCAT SafetySystem Funnels
SCAT™ FlexTubes
Solvent System
Unit Size:
Each
Product Code. 17337419
Brand: SymLine™ 106608
Only null left
Add to basket
Add to basket
Dovetail your SymLine™ System to suit your specific needs! Here, you will find a lot of useful ancillaries.
  • SymLine™ offers many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables to allow you to customize your SymLine™ system
  • All accessories are electrically dissipative and approved for use in laboratories with explosion hazards
Specifications
Closure Size GL25 (m) to GL25 (m)
Color Black
Diameter (Metric) Opening 10 mm
Includes Cabinet Feed-through, Counter Nut, Two Silicone Washers
For Use With (Application) SymLine™ System Customization
For Use With (Equipment) Cabinet (28 mm Borehole, 120 mm Wall Thickness)
Material HDPE-EC
Product Type Cabinet Feed-through