Description
- For use with accu-jet™ pro Pipette Controller.
- 100 to 240V
- 50 to 60Hz
Specifications
Specifications
| Type | AC Adapter |
| For Use With (Equipment) | accu-jet™ pro Pipette Controller |
| Plug Type | Continental Europe |
| Quantity | 1 Ea. |
| Electrical Requirements | 100/240 V, 50/60 Hz |
| Voltage | 100/240 V |
| Frequency | 50/60 Hz |