missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abrocitinib, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Abrocitinib |
| CAS | 1622902-68-4 |
| Molecular Formula | C14H21N5O2S |
| Quantity | 100 mg |
| Synonym | Cibinqo, N-[cis-3-[Methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]cyclobutyl]propane-1-sulfonamide, PF 04965842, PF 4965842, N-[cis-3-(Methyl-7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)cyclobutyl]-1-propanesulfonamide |
| InChI Formula | InChI=1S/C14H21N5O2S/c1-3-6-22(20,21)18-10-7-11(8-10)19(2)14-12-4-5-15-13(12)16-9-17-14/h4-5,9-11,18H,3,6-8H2,1-2H3,(H,15,16,17)/t10-,11+ |
| SMILES | CN([C@H]1C[C@H](C1)NS(CCC)(=O)=O)C2=C3C(NC=C3)=NC=N2 |
| IUPAC Name | N-((1s,3s)-3-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)cyclobutyl)propane-1-sulfonamide |
| Molecular Weight (g/mol) | 323.41 |
| Formula Weight | 323.1416 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction