missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SERPINA1 (Human) ELISA Kit
POA
Specifications
Detection Method | Colorimetric |
---|---|
Cross Reactivity | Human |
Product Type | ELISA Kit |
Regulatory Status | For research use only (RUO) |
Description
Enzyme immunoassay for the quantitative measurement of human SERPINA1.Specifications
Colorimetric | |
Human | |
ELISA Kit | |
For research use only (RUO) |