missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABLIM1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15776481 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15776481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15776481 Supplier Invitrogen™ Supplier No. PA558607

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RSSLLASRYD SPINSASHIP SSKTASLPGY GRNGLHRPVS TDFAQYNSYG DVSGGVRDYQ TLPDGHMPAM RMDR Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 91%.

This gene encodes a cytoskeletal LIM protein that binds to actin filaments via a domain that is homologous to erythrocyte dematin. LIM domains, found in over 60 proteins, play key roles in the regulation of developmental pathways. LIM domains also function as protein-binding interfaces, mediating specific protein-protein interactions. The protein encoded by this gene could mediate such interactions between actin filaments and cytoplasmic targets. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABLIM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABLIM1
Gene Accession No. O14639
Gene Alias 2210411C18Rik; 2610209L21Rik; 4833406P10Rik; 9330196J19Rik; ABLIM; ABLIM1; abLIM-1; actin binding LIM protein 1; actin-binding double zinc finger protein; actin-binding double-zinc-finger protein; actin-binding LIM protein 1; Actin-binding LIM protein family member 1; AV079770; AW060987; AW215784; KIAA0059; LIMAB1; Limatin; mKIAA0059; RGD1565768
Gene Symbols ABLIM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABLIM1. Recombinant protein control fragment (Product #RP-97029).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.