missing translation for 'onlineSavingsMsg'
Learn More

Abiraterone N-Oxide Sulfate Sodium Salt, TRC

Product Code. 30005531
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005531 1 mg 1mg
30004727 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005531 Supplier Toronto Research Chemicals Supplier No. TRCA1085201MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C24 H30 N O5 S . Na
Molecular Weight (g/mol) 467.5535
Synonym Abiraterone N-oxide sulfate sodium salt, (3β)-17-(1-Oxido-3-pyridinyl)androsta-5,16-dien-3-ol 3-(hydrogen sulfate) sodium salt, Abiraterone N-Oxide sulfate sodium, N-Oxide abiraterone sulfate sodium, Abiraterone sulfate N-oxide sodium salt
IUPAC Name sodium;[(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-(1-oxidopyridin-1-ium-3-yl)-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] sulfate
SMILES [Na+].C[C@]12CC[C@H]3[C@@H](CC=C4C[C@H](CC[C@]34C)OS(=O)(=O)[O-])[C@@H]1CC=C2c5ccc[n+]([O-])c5

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C24H31NO5S.Na/c1-23-11-9-18(30-31(27,28)29)14-17(23)5-6-19-21-8-7-20(16-4-3-13-25(26)15-16)24(21,2)12-10-22(19)23;/h3-5,7,13,15,18-19,21-22H,6,8-12,14H2,1-2H3,(H,27,28,29);/q;+1/p-1/t18-,19-,21-,22-,23-,24+;/m0./s1
Formula Weight 467.1742
Percent Purity >95%
Chemical Name or Material Abiraterone Sulfate N-Oxide Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.