missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCC11 Antibody (ABCC11/2438), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24276MFV450
This item is not returnable.
View return policy
Description
ABCC11 Monoclonal antibody specifically detects ABCC11 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ABCC11 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| ATP-binding cassette protein C11, ATP-binding cassette transporter MRP8, ATP-binding cassette transporter sub-family C member 11, ATP-binding cassette, sub-family C (CFTR/MRP), member 11, EWWD, MRP8ATP-binding cassette sub-family C member 11, Multidrug resistance-associated protein 8, multi-resistance protein 8, WW | |
| Recombinant fragment (around aa400-600) of human ABCC11 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| ABCC11/2438 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 85320 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction