missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAVR/KIAA0319L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
484.00 EUR
Specifications
| Antigen | KIAA0319L |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
AAVR/KIAA0319L Polyclonal specifically detects AAVR/KIAA0319L in Human samples. It is validated for Western Blot.Specifications
| KIAA0319L | |
| Polyclonal | |
| Rabbit | |
| Q8IZA0 | |
| 79932 | |
| Synthetic peptides corresponding to KIAA0319L(KIAA0319-like) The peptide sequence was selected from the middle region of KIAA0319L. Peptide sequence VTDTIGQQATAQVTVIVQPENNKPPQADAGPDKELTLPVDSTTLDGSKSS. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ44532, KIAA0319-like, KIAA1837dyslexia-associated protein KIAA0319-like protein, polycystic kidney disease 1-related | |
| KIAA0319L | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title