missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AARSD1 Recombinant Rabbit Monoclonal Antibody (012)
GREENER_CHOICE

Product Code. 16333124
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16333124 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16333124 Supplier Invitrogen™ Supplier No. MA528987

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human AARSD1.

AARSD1 functions in trans to edit the amino acid moiety from incorrectly charged tRNA(Ala).
TRUSTED_SUSTAINABILITY

Specifications

Antigen AARSD1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 012
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene AARSD1
Gene Accession No. Q9BTE6
Gene Alias 1110069E20Rik; 2310044P18Rik; AA589600; AARSD1; Alanyl-tRNA deacylase alaX; alanyl-tRNA editing protein Aarsd1; alanyl-tRNA synthetase domain containing 1; alanyl-tRNA synthetase domain-containing protein 1; Alax; AlaXp-II; MGC2744
Gene Symbols AARSD1
Host Species Rabbit
Immunogen Recombinant Human AARSD1 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.