missing translation for 'onlineSavingsMsg'
Learn More

AA26-9, MedChemExpress

Product Code. 16483537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483537 50 mg 50mg
16457518 10 mg 10mg
16449997 5 mg 5mg
16407476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483537 Supplier Medchem Express Supplier No. HY18522

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AA26-9 is a potent and broad spectrum serine hydrolase inhibitor. AA26-9 targets included serine peptidases, lipases, amidases, esterases, and thioesterases. AA26-9 shows inhibitory activity against approximately 1/3 of the 40+ serine hydrolases detected in immortalized T cell lines .

AA26-9 is a potent and broad spectrum serine hydrolase inhibitor. AA26-9 targets included serine peptidases, lipases, amidases, esterases, and thioesterases. AA26-9 shows inhibitory activity against approximately 1/3 of the 40+ serine hydrolases detected in immortalized T cell lines .

Specifications

Chemical Name or Material AA26-9
Physical Form Solid
CAS 1312782-34-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C7H10N4O
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 100 mg/mL (601.76 mM; Need ultrasonic)
SMILES O=C(N1N=NC=C1)N2CCCC2
Molecular Weight (g/mol) 166.18
Formula Weight 166.18
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.