Learn More
Honeywell™ A69 Peak Series Hard Hat
Description
- Rain trough design
- Selection of pin-lock and ratchet suspensions
- Size and headband height adjustable to ensure wearer comfort
- Slots to fit face shield and earmuffs
- Easy sweatband replacement system
- Personalizing with logo printing available
- ABS shell
Specifications
Specifications
| Color | White |
| Includes | Adjustable headband and suspension |
| Material | Shell: ABS, Suspension: Nylon web |
| Style | Ribbed, rain trough |
| Suspension Type | 6-point nylon web with ratchet adjustment |
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?