Learn More
Honeywell™ A69 Peak Series Hard Hat
Product Code. 15886795
Click to view available options
Suspension Type:
6-point nylon web with pin-lock adjustment
6-point nylon web with ratchet adjustment
Unit Size:
Each
Product Code. 15886795
Brand: Honeywell™ 933170
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Multipurpose helmet with ribbed ABS outer shell and 6-point nylon web suspension.
  • Rain trough design
  • Selection of pin-lock and ratchet suspensions
  • Size and headband height adjustable to ensure wearer comfort
  • Slots to fit face shield and earmuffs
  • Easy sweatband replacement system
  • Personalizing with logo printing available
  • ABS shell
TRUSTED_SUSTAINABILITY
Specifications
Color White
Includes Adjustable headband and suspension
Material Shell: ABS, Suspension: Nylon web
Style Ribbed, rain trough
Suspension Type 6-point nylon web with pin-lock adjustment