missing translation for 'onlineSavingsMsg'
Learn More

A-966492, MedChemExpress

Product Code. p-200098622
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486767

Brand: Medchem Express HY10614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1.5 nM, respectively.

A-966492 is a novel and potent inhibitor of PARP1 and PARP2 with Ki of 1 nM and 1.5 nM, respectively.

Specifications

Chemical Name or Material A-966492
CAS 934162-61-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H17FN4O
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (308.31 mM)
SMILES FC1=CC([C@H]2NCCC2)=CC=C1C3=NC4=C(C(N)=O)C=CC=C4N3
Molecular Weight (g/mol) 324.35
Formula Weight 324.35
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.