Learn More
Tocris Bioscience™ A 939572
Potent SCD-1 inhibitor
Brand: Tocris Bioscience™ 4845/10
This item is not returnable.
View return policy
Description
A 939572 is a potent stearoyl-CoA desaturase 1 (SCD1) inhibitor (IC50 = 0.4 nM). Selective for SCD1 over a range of kinases and hERG channels. Inhibits proliferation of squamous cell carcinoma FaDu cells in vitro. Also induces cell death of undifferentiated human embryonic stem cells (ESCs). Orally bioavailable.Specifications
| 4-(2-Chlorophenoxy)-N-[3-[(methylamino)carbonyl]phenyl]-1-piperidinecarboxamide | |
| 1032229-3-6 | |
| 10 mg |