Learn More
Fisherbrand™ 9mm Snap Cap, PE, Red with septum, for well plate micro inserts
Product Code. 10631623 Shop All Fisherbrand Products
Click to view available options
Color:
Red, White
Unit Size:
Pack of 960
Product Code. 10631623
Only null left
Add to basket
Add to basket
For use with well plate micro inserts.
  • Silicone/PTFE
  • White/Red
  • Slit
  • 1.9mm thickness
  • 45° shore A
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) Silicone/PTFE
Diameter (Metric) Opening 4 mm
Color Red, White
Diameter (Metric) Outer 9 mm
Material Polypropylene
Pre-slit Yes
For Use With (Application) GC, HPLC
Quantity 100
Thickness (Metric) 1.9 mm
Type Snap Cap