Learn More
Fisherbrand™ 9mm Short Thread Seal, Transparent, MS-Cap, thinned penetration area
Shop All Fisherbrand Products
Click to view available options
Unit Size:
Pack of 1000
Description
- Elimination of septum push-through
- Increased sealing capability
- Improved autosampler compatibility
- Flexibility to select the best septum for your instrument and applications
- Optimized ergonomics, fine texturing and evenly spaced ribbing for superior handling
- Easy-on, easy-off convenience with just one turn
- Pre-assembled caps and septa are convenient and minimize contamination from handling
- Closures have the profile of a crimp or snap closure for compatibility with robotic autosamplers
Specifications
Specifications
| Description | Short Thread Cap |
| Diameter (Metric) Outer | 9 mm |
| Septum | 100 |
| For Use With (Application) | GC, HPLC |
| Type | Screw Cap |