Learn More
Eppendorf™ 96-well V-Bottom PCR Microplates
GREENER_CHOICE
Product Code. 10248503 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Gray, White Bottom, White Bottom
White, Black Bottom, Black Bottom
White, Clear Bottom
Quantity:
240/Pk.
80/Pk.
Unit Size:
Pack of 240
Pack of 80
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
10248503 Gray, White Bottom, White Bottom 80/Pk. Pack of 80
10156933 White, Black Bottom, Black Bottom 240/Pk. Pack of 240
10136793 Gray, White Bottom, White Bottom 240/Pk. Pack of 240
10107223 White, Clear Bottom 80/Pk. Pack of 80
10146933 White, Black Bottom, Black Bottom 80/Pk. Pack of 80
10529203 White, Clear Bottom 240/Pk. Pack of 240
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10248503 Supplier Eppendorf™ Supplier No. 0030601670
Only null left
Specifications
Certified Yes
Format Standard
Detection Method Quantitative Protein Analysis
Array 8 x 12
Color Gray, White Bottom, White Bottom
Material Polypropylene
No. of Wells 96
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) For PCR techniques
Max. Heat Stability 121°C
Quantity 80/Pk.