Learn More
Labcon™ 96-place PCR Tube Storage Rack
Click to view available options
Color:
Green, Clear
Unit Size:
Pack of 10
Description
- Freezable to -80°C
- Made in USA
Specifications
Specifications
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Color | Green, Clear |
| For Use With (Equipment) | PCR Tubes |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur