Learn More
Uvex 95980 Insole For Uvex Naturform - Width 11
Product Code. 17279021 Shop All Uvex Products
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17279021
Brand: Uvex 9598044
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Comfortable climatic insole with antistatic properties for Uvex Performance (Naturform)
  • Full-surface foot shock absorption
  • Highly breathable and moisture-absorbent material
  • Shock absorption zones support natural foot movement
  • Skin compatibility and pleasant surface structure
  • Supports foot arch
  • Article No: 9598.0
  • Sizes: 38 to 48
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) Uvex Naturform
Material Microvelour
Size 44
Disposable Reusable
Gender Unisex
Fit Standard width
Latex Free Yes
Waterproof No