Learn More
Uvex 95980 Insole For Uvex Naturform - Width 11
Shop All Uvex Products
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- Full-surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No: 9598.0
- Sizes: 38 to 48
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | Uvex Naturform |
| Material | Microvelour |
| Size | 42 |
| Disposable | Reusable |
| Gender | Unisex |
| Fit | Standard width |
| Latex Free | Yes |
| Waterproof | No |