missing translation for 'onlineSavingsMsg'
Learn More

9-cis-Retinoic Acid, >98%, MP Biomedicals™

Product Code. 11409931 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409931 1 mg 1mg
11419931 5 mg 5mg
11429931 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409931 Supplier MP Biomedicals Supplier No. 0215872501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.44
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
ChEBI CHEBI:50648

Specifications

CAS 5300-03-8
Assay Percent Range ≥98%
Quantity 1 mg
Synonym alitretinoin, 9-cis-retinoic acid, panretin, 9-cis-tretinoin, panrexin, panretyn, panretin gel, toctino, 9-cis retinoic acid, 9 z-retinoic acid
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C
IUPAC Name (2E,4E,6Z,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
PubChem CID 449171
Formula Weight 300.44
Physical Form Powder
Chemical Name or Material 9-cis-Retinoic Acid, Cell Biology and Analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.