missing translation for 'onlineSavingsMsg'
Learn More

9-Bromophenanthrene-d9, TRC

Product Code. 30342909
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342909 2.5 mg 2.5mg
30329001 10 mg 10mg
30333608 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30342909 Supplier Toronto Research Chemicals Supplier No. TRCB6872022.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-Bromophenanthrene-d9
CAS 2055815-26-2
Molecular Formula C14D9Br
Quantity 2.5 mg
Synonym 9-Phenanthryl Bromide-d9, NSC 400708-d9
InChI Formula InChI=1S/C14H9Br/c15-14-9-10-5-1-2-6-11(10)12-7-3-4-8-13(12)14/h1-9H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
SMILES BrC1=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2C3=C1C([2H])=C([2H])C([2H])=C3[2H]
IUPAC Name 9-bromophenanthrene-1,2,3,4,5,6,7,8,10-d9
Molecular Weight (g/mol) 266.18
Formula Weight 265.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.