Learn More
9-Aminominocycline Hydrochloride (~90%), TRC
Specifications
Specifications
| Chemical Name or Material | (4S,4aS,5aR,12aS)-9-Amino-4,7-bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride |
| CAS | 149934-21-4 |
| Molecular Formula | C23H29ClN4O7 |
| Quantity | 100 mg |
| Synonym | 2-Naphthacenecarboxamide, 9-amino-4,7-bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-, hydrochloride (1:1), (4S,4aS,5aR,12aS)-, 2-Naphthacenecarboxamide, 9-amino-4,7-bis(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,10,12,12a-tetrahydroxy-1,11-dioxo-, monohydrochloride, [4S-(4α,4aα,5aα,12aα)]- (9CI), 9-Aminominocycline hydrochloride |
| InChI Formula | InChI=1S/C23H28N4O7.ClH/c1-26(2)12-7-11(24)17(28)14-9(12)5-8-6-10-16(27(3)4)19(30)15(22(25)33)21(32)23(10,34)20(31)13(8)18(14)29;/h7-8,10,16,28,30-31,34H,5-6,24H2,1-4H3,(H2,25,33);1H/t8-,10-,16-,23-;/m0./s1 |
| SMILES | Cl.CN(C)[C@H]1[C@@H]2C[C@@H]3Cc4c(cc(N)c(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)N(C)C |
| IUPAC Name | (4S,4aS,5aR,12aS)-9-amino-4,7-bis(dimethylamino)-3,10,12,12a-tetrahydroxy-1,11-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 508.9 |
| Formula Weight | 508.1725 |