Learn More
9,9-Di-n-octylfluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Product Code. p-7034283
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439465 1 g 1g
15449465 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15439465 Supplier Thermo Scientific Alfa Aesar Supplier No. H51953.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 302554-81-0
Molecular Formula C35H53BO2
Molecular Weight (g/mol) 516.617
MDL Number MFCD16294553
InChI Key VOASJDUTCQKQLE-UHFFFAOYSA-N
Synonym 2-9,9-dioctyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-di-n-octylfluorene-2-boronic acid pinacol ester, 2-9,9-dioctyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 3-hydroxy-2,3-dimethylbutan-2-yl hydrogen 9,9-dioctyl-9h-fluoren-2-ylboronate, amtb255, 2-9,9-dioctylfluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dioctylfluorene-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolyl-9,9-dioctylfluorene, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dioctylfluorene, 2-9,9-di-n-octyl-2-fluorenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11477856
IUPAC Name 2-(9,9-dioctylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3(CCCCCCCC)CCCCCCCC
Specifications
Quantity 1 g
Beilstein 8661191
Formula Weight 516.61
Percent Purity 95%
Chemical Name or Material 9,9-Di-n-octylfluorene-2-boronic acid pinacol ester