Learn More
Biochrom 8 mm Pozi Head Screw (K22) Spare Part, for UV-Vis Spectrophotometers
Description
- Ready to use
- Pack of 20
Specifications
Specifications
| Description | 8 mm Pozi Head Screw (K22) Spare Part, Pk. of 20, for UV-Vis Spectrophotometers |
| Diameter (English) | 0.3 in. |
| Diameter (Metric) | 8 mm |
| Disposable | Reusable |
| For Use With (Equipment) | UV-Visible Spectrophotometers |
| Shape | Cylindrical |
For research use only.