Learn More
8-Oxo-Benzylguanine, TRC
Product Code. 30421397
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421397 500 mg
30421909 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30421397 Supplier Toronto Research Chemicals Supplier No. TRCO847390500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 158754-46-2
Molecular Formula C12H11N5O2
Molecular Weight (g/mol) 257.25
Synonym 2-Amino-7,9-dihydro-6-(phenylmethoxy)-8H-purin-8-one, NU 6043
IUPAC Name 2-amino-6-phenylmethoxy-7,9-dihydropurin-8-one
SMILES C1=CC=C(C=C1)COC2=NC(=NC3=C2NC(=O)N3)N
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C12H11N5O2/c13-11-15-9-8(14-12(18)16-9)10(17-11)19-6-7-4-2-1-3-5-7/h1-5H,6H2,(H4,13,14,15,16,17,18)
Formula Weight 257.0913
Percent Purity >95%
Chemical Name or Material 8-Oxo-Benzylguanine
Grade HPLC