missing translation for 'onlineSavingsMsg'
Learn More

8-Oxo-2’-deoxyguanosine, TRC

Product Code. 30007884
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007884 1 mg 1mg
30012601 5 mg 5mg
30011283 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007884 Supplier Toronto Research Chemicals Supplier No. TRCO8502501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88847-89-6
Molecular Formula C10 H13 N5 O5
Molecular Weight (g/mol) 283.24
Synonym Guanosine, 2'-deoxy-7,8-dihydro-8-oxo-, 2'-Deoxy-7,8-dihydro-8-oxoguanosine, 7,8-Dihydro-8-oxo-2'-deoxyguanosine, 7,8-Dihydro-8-oxodeoxyguanosine, 8-Hydroxy-2'-deoxyguanosine, 8-Hydroxydeoxyguanosine, 8-Oxo-2'-deoxyguanosine, 8-Oxo-7,8-dihydro-2'-deoxyguanosine, 8-Oxo-7,8-dihydrodeoxyguanosine, 8-Oxo-dG, 8-Oxodeoxyguanosine
IUPAC Name 2-amino-8-hydroxy-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one
SMILES NC1=Nc2c(nc(O)n2[C@H]3C[C@H](O)[C@@H](CO)O3)C(=O)N1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C10H13N5O5/c11-9-13-7-6(8(18)14-9)12-10(19)15(7)5-1-3(17)4(2-16)20-5/h3-5,16-17H,1-2H2,(H,12,19)(H3,11,13,14,18)/t3-,4+,5+/m0/s1
Formula Weight 283.0917
Percent Purity >95%
Chemical Name or Material 8-Hydroxy-2'-deoxyguanosine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.