missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 8-OHdG Competitive ELISA Kit

Product Code. 17951092
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 17951092

Brand: Invitrogen™ EEL004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Oxygen is necessary for aerobic life, it can also participate in potentially toxic reactions involving oxygen free radicals and transition metals such as iron that damage membranes, proteins, and nucleic acids. n a normal human cell, there is a steady accumulation of DNA lesions with time. Substantial parts of these lesions are due to endogenous factors that damage DNA. These include reactive oxygens pecies derived from oxidative respiration. Additionally, malignant cells can produce hydrogen peroxide at levels as high as those characteristics for stimulated polymorphonuclear leukocytes. Some tumors may stimulate the defense systems of the body so that they react against the tumor to produce cytokines. Some cytokines can produce large amounts of ROS. 8-OHdG is one of the most widely studied lesions of this type. The presenceof 8-OHdG residues in DNA leads to GC to TA transversion unless repairs are made before DNA replication. Therefore, the presence of 8-OHdG may lead to mutagenesis. Furthermore, many observations indicate a direct correlation between 8-OHdG formation and carcinogenesis in vivo. It is generally accepted that the repair products of cellular DNA, for example 8-OHdG, are excreted into the urine without further metabolism. Therefore, 8-OHdG has been proposed as a urinary biomarker for in vivo oxidative DNA lesions.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.56 to 100 ng/mL
Assay Sensitivity 0.94 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target 8-OHdG
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.