Learn More
8-Hydroxy-7-iodoquinoline-5-sulfonic Acid, TRC
Product Code. 30406461
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406461 1 g 1g
30406903 500 mg -
30400943 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30406461 Supplier Toronto Research Chemicals Supplier No. TRCH9436781G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 547-91-1
Molecular Formula C9H6NO4SI
Molecular Weight (g/mol) 351.11
Synonym 8-Hydroxy-7-iodo-5-quinolinesulfonic Acid, 5-Sulfo-7-iodo-8-hydroxyquinoline, 5-Sulfo-7-iodo-8-quinolinol, 7-Iodo-5-sulfonic acid-8-hydroxyquinoline, 7-Iodo-8-hydroxylquinoline-5-sulfonic Acid, 7-Iodo-8-hydroxyquinoline-5-sulfonic Acid, 7-Iodo-8-quinolinol-5-sulfonic Acid, 7-Iodooxine-5-sulfonic Acid, 8-Hydroxy-7-iodoquinoline-5-sulfonic Acid, Ferron, Ferron (analytical reagent), Loretin, Meditrene, NSC 3784, Quiniophen, Yellon
IUPAC Name 8-hydroxy-7-iodoquinoline-5-sulfonic acid
SMILES Oc1c(I)cc(c2cccnc12)S(=O)(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H6INO4S/c10-6-4-7(16(13,14)15)5-2-1-3-11-8(5)9(6)12/h1-4,12H,(H,13,14,15)
Formula Weight 350.9062
Chemical Name or Material 8-Hydroxy-7-iodoquinoline-5-sulfonic Acid