Learn More
8-Chloro-1-octanol, 98%
Product Code. 11492040
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492040 5 mL 5mL
11402050 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11492040 Supplier Thermo Scientific Alfa Aesar Supplier No. H28417.AA
Request Bulk or Custom Format
CAS: 23144-52-7 | C8H17ClO | 164.673 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23144-52-7
Molecular Formula C8H17ClO
Molecular Weight (g/mol) 164.673
MDL Number MFCD00040005
InChI Key YDFAJMDFCCJZSI-UHFFFAOYSA-N
Synonym 8-chloro-1-octanol, 1-octanol, 8-chloro, 8-chlorooctanol, 8-chloro-octan-1-ol, 8-chloro-1-n-octanol, 8-chlor-1-octanol, 8-chlor-octan-1-ol, acmc-1cr4x
PubChem CID 90012
IUPAC Name 8-chlorooctan-1-ol
SMILES C(CCCCCl)CCCO
Specifications
Density 0.976
Boiling Point 129°C to 130°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.458
Quantity 5 mL
Formula Weight 164.67
Percent Purity 98%
Chemical Name or Material 8-Chloro-1-octanol