missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromo-cAMP sodium salt, MedChemExpress
Description
8-Bromo-cAMP sodium salt (8-Br-Camp sodium salt), a cyclic AMP analog, is an activator of cyclic AMP-dependent protein kinase (PKA).
Specifications
Specifications
| Chemical Name or Material | 8-Bromo-cAMP sodium salt |
| CAS | 76939-46-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C10H10BrN5NaO6P |
| Quantity | 100 mg |
| Synonym | 8-Br-Camp sodium salt |
| Solubility Information | DMSO : 250 mg/mL (581.29 mM; Need ultrasonic) ∣H2O : 100 mg/mL (232.51 mM; Need ultrasonic) |
| SMILES | BrC1=NC2=C(N=CN=C2N)N1[C@H](O[C@@](CO3)([H])[C@@]4([H])OP3(O[Na])=O)[C@@H]4O |
| Molecular Weight (g/mol) | 430.08 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?