Learn More
(7S,9S)-9-Acetyl-6,7,9,11-tetrahydroxy-7,8,9,10-tetrahydro-5,12-naphthacenedione, TRC
Specifications
Specifications
| Chemical Name or Material | (7S,9S)-9-Acetyl-6,7,9,11-tetrahydroxy-7,8,9,10-tetrahydro-5,12-naphthacenedione |
| CAS | 60660-75-5 |
| Molecular Formula | C20 H16 O7 |
| Quantity | 10 mg |
| Synonym | 5,12-Naphthacenedione, 9-acetyl-7,8,9,10-tetrahydro-6,7,9,11-tetrahydroxy-, (7S-cis)-, (7S,9S)-9-Acetyl-7,8,9,10-tetrahydro-6,7,9,11-tetrahydroxy-5,12-naphthacenedione, (+)-4-Demethoxydaunomycinone, (+)-Idarubicinone, 4-Demethoxydaunomycinone, Idarubicin aglycone, Idarubicinone |
| InChI Formula | InChI=1S/C20H16O7/c1-8(21)20(27)6-11-13(12(22)7-20)19(26)15-14(18(11)25)16(23)9-4-2-3-5-10(9)17(15)24/h2-5,12,22,25-27H,6-7H2,1H3/t12-,20-/m0/s1 |
| SMILES | CC(=O)[C@@]1(O)C[C@H](O)c2c(O)c3C(=O)c4ccccc4C(=O)c3c(O)c2C1 |
| IUPAC Name | (7S,9S)-9-acetyl-6,7,9,11-tetrahydroxy-8,10-dihydro-7H-tetracene-5,12-dione |
| Molecular Weight (g/mol) | 368.34 |
| Formula Weight | 368.0896 |