Learn More
Brady™ 7950 Series Thermal Transfer Printer Ribbons
Product Code. 11528824
Click to view available options
Color:
Black
Blue
Red
Dimensions (L x W):
300 m x 60 mm
70 m x 110 mm
70 m x 35 mm
70 m x 65 mm
70 m x 85 mm
Diameter (Metric):
12.7 mm I.D.
25.4 mm I.D.
Unit Size:
Each
Product Code. 11528824
Brand: Brady™ 804468
Only null left
Add to basket
Add to basket
Provides strong environmental resistance against smearing and chemicals.
7950 Series Thermal Transfer Printer Ribbons
  • Wax/resin formulation
  • Ink side out ribbon
Specifications
Product Type Thermal Transfer Printer Ribbon
Color Black
Length (Metric) 70 m
Diameter (Metric) 12.7 mm I.D.
Dimensions (L x W) 70 m x 85 mm
Description Black 7950 Series Thermal Transfer Printer Ribbon
Printable Sides Ink side out
Alternate Part Number R7950-85X70/O