Learn More
Applied Biosystems™ 7900HT System Standard Upgrade Kits
Product Code. 15893354 Shop All Applied Biosystems Products
Click to view available options
Description:
384-well Kit
96-well Kit
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 15893354
Brand: Applied Biosystems™ 4331405
Optional accessory for the 7900HT Fast RT-PCR System, which is available is either a 96- or 384-well block format.
The interchangeable 96- and 384-well block capability combined with the optional Automation Accessory offer unmatched flexibility. These features make the 7900HT system ideally suited to meet the high-throughput requirements of today’s drug discovery process. Key applications include gene expression quantitation and the detection of single nucleotide polymorphisms (SNPs) using the fluorogenic 5' nuclease assay.The 7900HT Fast Real-Time PCR System is the only real-time quantitative PCR system that combines 96- and 384-well plate compatibility and the TaqMan™ Low Density Array with fully automated robotic loading-and now also offers optional Fast real-time PCR capability.
4331405, 96-well block
Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
4331406, 384-well block
Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 7900HT System
Product Type Block Upgrade Kit
For Use With (Application) Real Time PCR (qPCR), Real Time PCR-based Gene Expression Profiling
Description 96-well Kit
Quantity 1 Kit