Learn More
75 cSt Mineral Oil Blank - Trace metals and sulfur reported, VHG™
Description
75 cSt Mineral Oil Blank - Trace metals and sulfur reported
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Arsenic/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Copper/Iron/Lanthanum/Lead/Lithium/Magnesium/Manganese/Mercury/Molybdenum/Nickel/Phosphorus/Potassium/Scandium/Selenium/Silicon/Silver/Sodium/Strontium/Sulfur/Thallium/Tin/Titanium/Vanadium/Yttrium/Zinc |
| CAS | 7429-90-5, 7440-36-0, 7440-38-2, 7440-39-3, 7440-69-9, 7440-42-8 |
| Analyte or Component Names | Aluminium/Antimony/Arsenic/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Cobalt/Copper/Iron/Lanthanum/Lead/Lithium/Magnesium/Manganese/Mercury/Molybdenum/Nickel/Phosphorus/Potassium/Scandium/Selenium/Silicon/Silver/Sodium/Strontium/Sulfur/Thallium/Tin/Titanium/Vanadium/Yttrium/Zinc |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | 75 cSt Mineral Oil |
| Quantity | 3.78 L |
| Concentration or Composition (by Analyte or Components) | Aluminium <1 μg/g/Antimony <1 μg/g/Arsenic <1 μg/g/Barium <1 μg/g/Bismuth <1 μg/g/Boron <1 μg/g/Cadmium <1 μg/g/Calcium <1 μg/g/Chromium <1 μg/g/Cobalt <1 μg/g/Copper <1 μg/g/Iron <1 μg/g/Lanthanum <1 μg/g/Lead <1 μg/g/Lithium <1 μg/g/Magnesium <1 μg/g/Manganese <1 μg/g/Mercury <1 μg/g/Molybdenum <1 μg/g/Nickel <1 μg/g/Phosphorus <1 μg/g/Potassium <1 μg/g/Scandium <1 μg/g/Selenium <1 μg/g/Silicon <1 μg/g/Silver <1 μg/g/Sodium <1 μg/g/Strontium <1 μg/g/Sulfur <1 μg/g/Thallium <1 μg/g/Tin <1 μg/g/Titanium <1 μg/g/Vanadium <1 μg/g/Yttrium <1 μg/g/Zinc <1 μg/g |