Learn More
75 cSt Mineral Oil Blank - Trace metals reported, VHG™
Description
75 cSt Mineral Oil Blank - Trace metals reported
Specifications
Specifications
| Chemical Name or Material | 75 cSt Mineral Oil Blank |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | 75 cSt Mineral Oil Blank |
| Shipping Condition | Room Temperature |
| IUPAC Name | 75 cSt Mineral Oil Blank |
| Solvent or Matrix | 75 cSt Hydrocarbon Oil |
| Quantity | 1 gal |