Learn More
Applied Biosystems™ 7300 Real Time PCR Systems Spectral Calibration Kit
GREENER_CHOICE
Product Code. 10617705 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10617705 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10617705 Supplier Applied Biosystems™ Supplier No. 4349182
Easy-to-use, preloaded, 96-well optical reaction plates enable quick and easy calibration of the 7300 Real-Time PCR System.
This easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™/SYBR™ Green I, VIC™/JOE™, and NED™/TAMRA™/ROX™ dyes on the Applied Biosystems™ 7300 Real-Time PCR System. The Applied Biosystems™ 7300 Real-Time PCR Systems Spectral Calibration Kit I contains one background plate, one region of interest (ROI) calibration plate, and seven spectral calibration plates with seven separate dye standards.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Contains one background plate, one ROI calibration plate,and seven spectral calibration plates with FAM™⁄SYBR™ Green I, VIC™⁄JOE™, and NED™⁄TAMRA™⁄ROX™ dye standards. Store at -15 to -25°C. Protect from light.
Calibrated Dye FAM, JOE, NED, ROX, TAMRA™, VIC, SYBR™ Green
For Use With (Equipment) 7300 System
Detection Method Fluorescence
Product Type Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 Kit
Calibration Type Spectral Calibration