Learn More
Hamilton™ 7000 Series Knurled Hub Syringes
Product Code. p-8543020
Click to view available options
Volume (Metric):
1 μL
Needle Gauge:
23 G
25 G
Unit Size:
Each
Product Code. 15153038
Brand: Hamilton™ 80108
Modified Microliter syringes with a plunger-in-needle design.
  • The only zero dead volume syringe available
  • Fine gauge tungsten plunger wire extends to the tip of the needle
  • Ensures that all solvent is expelled
Specifications
Autoclavable Autoclavable
Needle Gauge 25 G
Diameter (Metric) Inner Needle 0.145 mm
Diameter (Metric) Outer 7.82 mm
Diameter (Metric) Outer Needle 0.51 mm
Product Type Syringe Needle
Max. Pressure 413.7 bar
Max. Temperature (Metric) 50°C
Model 7001
Quantity 1/Pk.
Volume (Metric) 1 μL
Needle Point Style Custom
Connection Type Knurled Hub