Learn More
Hamilton™ 700 Series Microliter™ Carbon Analyzer Syringes
Product Code. 11582475
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
Needle Gauge:
24 G
26 G
Product Type:
Carbon Analyzer Syringe
Microliter Syringe
Unit Size:
Each
Product Code. 11582475
Brand: Hamilton™ 80660
Syringes are designed for water analysis with Total Organic Carbon (TOC)
  • Microliter syringes with a stainless-steel plunger that is hand-fitted to the glass barrel and designed for liquid sampling
  • Syringes have a carbon analyzer (CA) termination
  • Available in a 50 or 250μL volume
Specifications
Autoclavable Not Autoclavable
Material Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Needle Gauge 24 G
Diameter (English) Inner 0.061 in.
Diameter (English) Inner Needle 0.012 in.
Diameter (Metric) Inner Needle 0.31 mm
Diameter (English) Outer Needle 0.022 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 1.46 mm
Diameter (Metric) Outer 7.75 mm
Diameter (Metric) Outer Needle 0.57 mm
For Use With (Application) Designed for water analysis with total organic carbon
Length (English) 2 in.
Length (English) Needle 2 in.
Length (Metric) 51 mm
Product Type Microliter Syringe
Max. Pressure 2000 psi
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 710
Needle Point Number 3
Min. Temperature (Metric) 10°C
Quantity Ea.
Volume (Metric) 100 μL
Needle Point Style Blunt
Connection Type Carbon Analyzer, Cemented