missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(tert-Butyldimethylsilyl)oxy Cannflavin A Pivalate, TRC
Specifications
Specifications
| Chemical Name or Material | 7-(tert-Butyldimethylsilyl)oxy Cannflavin A Pivalate |
| Analyte or Component Names | 7-(tert-Butyldimethylsilyl)oxy Cannflavin A Pivalate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C37 H50 O7 Si |
| Synonym | (E)-4-(7-((tert-Butyldimethylsilyl)oxy)-6-(3,7-dimethylocta-2,6-dien-1-yl)-5-hydroxy-4-oxo-4H-chromen-2-yl)-2-methoxyphenyl Pivalate |
| InChI Formula | InChI=1S/C37H50O7Si/c1-23(2)14-13-15-24(3)16-18-26-30(44-45(11,12)37(7,8)9)22-32-33(34(26)39)27(38)21-29(42-32)25-17-19-28(31(20-25)41-10)43-35(40)36(4,5)6/h14,16-17,19-22,39H,13,15,18H2,1-12H3/b24-16+ |
| Solution Type | Neat |
| SMILES | COc1cc(ccc1OC(=O)C(C)(C)C)C2=CC(=O)c3c(O)c(C\C=C(/C)\CCC=C(C)C)c(O[Si](C)(C)C(C)(C)C)cc3O2 |
| IUPAC Name | [4-[7-[tert-butyl(dimethyl)silyl]oxy-6-[(2E)-3,7-dimethylocta-2,6-dienyl]-5-hydroxy-4-oxochromen-2-yl]-2-methoxyphenyl] 2,2-dimethylpropanoate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?