Learn More
7-O-(Triethylsilyl)baccatin III, Mikromol
Description
7-O-(Triethylsilyl)baccatin III, Mikromol
Specifications
Specifications
| Chemical Name or Material | 7-O-(Triethylsilyl) Baccatin III |
| CAS | 115437-21-3 |
| Analyte or Component Names | 7-O-(Triethylsilyl) Baccatin III |
| Shipping Condition | Room Temperature |
| Molecular Formula | C37 H52 O11 Si |
| Certifications/Compliance | ISO 9001 |
| Synonym | 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-9,11-dihydroxy-4a,8,13,13-tetramethyl-4-[(triethylsilyl)oxy]-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-, 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-9,11-dihydroxy-4a,8,13,13-tetramethyl-4-[(triethylsilyl)oxy]-, [2aR-(2aα,4β,4aβ,6β,9α,11α,12α,12aα,12bα)]-, 7-(Triethylsilyl)baccatin III, 7-O-(Triethylsilyl)baccatin III |
| InChI Formula | InChI=1S/C37H52O11Si/c1-10-49(11-2,12-3)48-26-18-27-36(20-44-27,47-23(6)39)30-32(46-33(42)24-16-14-13-15-17-24)37(43)19-25(40)21(4)28(34(37,7)8)29(45-22(5)38)31(41)35(26,30)9/h13-17,25-27,29-30,32,40,43H,10-12,18-20H2,1-9H3/t25-,26-,27+,29+,30-,32-,35+,36-,37+/m0/s1 |
| Solution Type | Neat |
| SMILES | CC[Si](CC)(CC)O[C@H]1C[C@H]2OC[C@@]2(OC(=O)C)[C@H]3[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](O)C(=C([C@@H](OC(=O)C)C(=O)[C@]13C)C5(C)C)C |
| Show More |