missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-O-(Triethylsilyl)-10-deacetylbaccatin III, Mikromol
Description
7-O-(Triethylsilyl)-10-deacetylbaccatin III, Mikromol
Specifications
Specifications
| Chemical Name or Material | 7-O-(Triethylsilyl)-10-deacetyl Baccatin III |
| CAS | 115437-18-8 |
| Analyte or Component Names | 7-O-(Triethylsilyl)-10-deacetyl Baccatin III |
| Shipping Condition | Room Temperature |
| Molecular Formula | C35 H50 O10 Si |
| Certifications/Compliance | ISO 9001 |
| Synonym | 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 12b-(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-6,9,11-trihydroxy-4a,8,13,13-tetramethyl-4-[(triethylsilyl)oxy]-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-, 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 12b-(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-6,9,11-trihydroxy-4a,8,13,13-tetramethyl-4-[(triethylsilyl)oxy]-, [2aR-(2aα,4β,4aβ,6β,9α,11α,12α,12aα,12bα)]-, 10-Deacetyl-7-O-(triethylsilyl)baccatin III, 10-Deacetyl-7-triethylsilylbaccatin III, 7-(Triethylsilyl)-10-deacetylbaccatin III, 7-O-(Triethylsilyl)-10-deacetyl baccatin III, 7-O-Triethylsilyl-10-desacetylbaccatin III |
| InChI Formula | InChI=1S/C35H50O10Si/c1-9-46(10-2,11-3)45-24-17-25-34(19-42-25,44-21(5)36)28-30(43-31(40)22-15-13-12-14-16-22)35(41)18-23(37)20(4)26(32(35,6)7)27(38)29(39)33(24,28)8/h12-16,23-25,27-28,30,37-38,41H,9-11,17-19H2,1-8H3/t23-,24-,25+,27+,28-,30-,33+,34-,35+/m0/s1 |
| Solution Type | Neat |
| SMILES | CC[Si](CC)(CC)O[C@H]1C[C@H]2OC[C@@]2(OC(=O)C)[C@H]3[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](O)C(=C([C@@H](O)C(=O)[C@]13C)C5(C)C)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?