Learn More
6H05 (TFA), MedChemExpress
Product Code. 16467867
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16467867
Brand: Medchem Express HY12408A
MedChemExpress 6H05 TFA is a selective, and allosteric inhibitor of oncogenic mutant K-Ras(G12C).
6H05 TFA is a selective, and allosteric inhibitor of oncogenic mutant K-Ras(G12C).
Specifications
Chemical Name or Material 6H05 (TFA)
CAS 2061344-88-3
Color White
Purity Grade Notes Research
Molecular Formula C22H31ClF3N3O4S3
Quantity 5 mg
Solubility Information DMSO : ≥ 51 mg/mL (86.42 mM)
SMILES O=C(C1CCN(C(CSC2=CC=C(Cl)C=C2)=O)CC1)NCCSSCCN(C)C.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 590.14
Formula Weight 590.14
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease