missing translation for 'onlineSavingsMsg'
Learn More

6H05 (TFA), MedChemExpress

Product Code. 16435148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. Quantity unitSize
16435148 10 mg 10mg
16467867 5 mg 5mg
2 options
This item is not returnable. View return policy

Product Code. 16435148

Brand: Medchem Express HY12408A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 6H05 TFA is a selective, and allosteric inhibitor of oncogenic mutant K-Ras(G12C).

6H05 TFA is a selective, and allosteric inhibitor of oncogenic mutant K-Ras(G12C).

Specifications

Chemical Name or Material 6H05 (TFA)
CAS 2061344-88-3
Color White
Purity Grade Notes Research
Molecular Formula C22H31ClF3N3O4S3
Quantity 10 mg
Solubility Information DMSO : ≥ 51 mg/mL (86.42 mM)
SMILES O=C(C1CCN(C(CSC2=CC=C(Cl)C=C2)=O)CC1)NCCSSCCN(C)C.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 590.14
Formula Weight 590.14
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.