missing translation for 'onlineSavingsMsg'
Learn More

6-Nitropyridine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-7032950
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15437577 250 mg 250mg
15427577 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15437577

Brand: Thermo Scientific Alfa Aesar H35766.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26893-68-5
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD04114205
InChI Key MIHFOFLNJIPDRY-UHFFFAOYSA-N
Synonym 6-nitropyridine-2-carboxylic acid, 6-nitropicolinic acid, 6-nitro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 6-nitro, picolinic acid, 6-nitro, 2-carboxy-6-nitropyridine, 2-pyridinecarboxylicacid, 6-nitro
PubChem CID 578841
SMILES C1=CC(=NC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 170°C to 173°C
Quantity 250 mg
Formula Weight 168.11
Percent Purity 97%
Chemical Name or Material 6-Nitropyridine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.