missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Product Code. p-7054046
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15408508

Brand: Thermo Scientific Alfa Aesar H61911.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloroindole-3-carboxaldehyde can react with dimethyldisulfane to get 6-chloro-(3-thiomethyl)-indole.

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 703-82-2
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD06657152
InChI Key CTNIXLBHXMSZKL-UHFFFAOYSA-N
Synonym 6-chloroindole-3-carboxaldehyde, 6-chloro indole-3-carbaldehyde, 6-chloro-3-formylindole, 6-chloro-3-indolecarboxaldehyde, 1h-indole-3-carboxaldehyde, 6-chloro, 6-chloro-1h-indole-3-carboxaldehyde, pubchem7693, acmc-209oec, ksc494c8d, 6-chloroindole-3-carbaldehyde
PubChem CID 12614669
IUPAC Name 6-chloro-1H-indole-3-carbaldehyde
SMILES C1=CC2=C(C=C1Cl)NC=C2C=O

Specifications

Color Brown
Quantity 5 g
Solubility Information Slightly soluble in water
Formula Weight 179.6
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Chloroindole-3-carboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.