Learn More
6-Chloro-7-iodo-7-deazapurine, 96%
Product Code. 15458007
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15458007
Brand: Thermo Scientific Alfa Aesar H51128.03
Request Bulk or Custom Format
CAS: 123148-78-7 | C6H3ClIN3 | 279.465 g/mol
6-Chloro-7-iodo-7-deazapurine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Chloro-7-iodo-7-deazapurine is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123148-78-7
Molecular Formula C6H3ClIN3
Molecular Weight (g/mol) 279.465
MDL Number MFCD09263258
InChI Key CBWBJFJMNBPWAL-UHFFFAOYSA-N
Synonym 4-chloro-5-iodo-7h-pyrrolo 2,3-d pyrimidine, 6-chloro-7-iodo-7-deazapurine, 4-chloro-5-iodo-7h-pyrrol 2,3-d pyrimidine, 4-chloro-5-iodo-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodopyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro-5-iodo, acmc-20agy2, amtns039, 6-chloro-7-iod-7-deazapurine
PubChem CID 14809281
IUPAC Name 4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=C(C2=C(N1)N=CN=C2Cl)I
Specifications
Melting Point ∽180°C (decomposition)
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 279.47
Percent Purity 96%
Chemical Name or Material 6-Chloro-7-iodo-7-deazapurine