missing translation for 'onlineSavingsMsg'
Learn More

6-Aminohexanoic Acid Hydrochloride, TRC

Product Code. 30314072
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314072 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314072 Supplier Toronto Research Chemicals Supplier No. TRCA6030151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Aminohexanoic Acid Hydrochloride
CAS 4321-58-8
Molecular Formula C6 H13 N O2 . H Cl
Quantity 1 g
Synonym ε-Aminocaproic Acid Hydrochloride, 6-Aminocaproic Acid Hydrochloride
InChI Formula InChI=1S/C6H13NO2.ClH/c7-5-3-1-2-4-6(8)9;/h1-5,7H2,(H,8,9);1H
SMILES Cl.NCCCCCC(=O)O
IUPAC Name 6-aminohexanoic acid;hydrochloride
Molecular Weight (g/mol) 167.634
Formula Weight 167.0713
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.