missing translation for 'onlineSavingsMsg'
Learn More

5Beta-Cholestan-3Alpha,7Alpha,12Alpha,23R,25-pentol, TRC

Product Code. 30134877
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
10 mg
Unit Size:
0.5mg
10mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134877 10 mg 10mg
30136356 2.5 mg 2.50mg
30010144 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134877 Supplier Toronto Research Chemicals Supplier No. TRCC43171510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59906-14-8
Molecular Formula C27 H48 O5
Molecular Weight (g/mol) 452.67
Synonym 5β-Cholestan-3α,7α,12α,23R,25-pentol
IUPAC Name (3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-17-[(1R,3R)-3,5-dihydroxy-1,5-dimethyl-hexyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-3,7,12-triol
SMILES C[C@H](C[C@@H](O)CC(C)(C)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H48O5/c1-15(10-18(29)14-25(2,3)32)19-6-7-20-24-21(13-23(31)27(19,20)5)26(4)9-8-17(28)11-16(26)12-22(24)30/h15-24,28-32H,6-14H2,1-5H3/t15-,16+,17-,18-,19-,20+,21+,22-,23+,24+,26+,27-/m1/s1
Formula Weight 452.3502
Chemical Name or Material 5Beta-Cholestan-3Alpha,7Alpha,12Alpha,23R,25-pentol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.