missing translation for 'onlineSavingsMsg'
Learn More

5alpha-Cholestan-3beta-ol, TRC

Product Code. 30138135
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138135 10 g 10g
30009885 100 mg 100mg
30010021 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138135 Supplier Toronto Research Chemicals Supplier No. TRCC43171010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-97-7
Formula Weight 388.3705
InChI Formula InChI=1S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28H,6-17H2,1-5H3/t19-,20+,21+,22+,23-,24+,25+,26+,27-/m1/s1
IUPAC Name (3S,5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molecular Formula C27 H48 O
Molecular Weight (g/mol) 388.67
Percent Purity >95%
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Synonym Cholestan-3-ol, (3β,5α)-, 5α-Cholestan-3β-ol (8CI), (3β,5α)-Cholestan-3-ol, 3β-Hydroxy-5α-cholestane, 3β-Hydroxycholestane, 5α(H)-Cholestan-3β-ol, 5α-Cholestanol, 5α-Dihydrocholesterol, Cholestan-3β-ol, Cholestanol, Dihydrocholesterin, Dihydrocholesterol, Dihydrolathosterol, Lathostanol, NSC 18188, Zymostanol, β-Cholestanol
Chemical Name or Material 5alpha-Cholestan-3-beta-ol
Grade HPLC
Show More Show Less

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.